missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexamethasone Sodium Phosphate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Dexamethasone disodium phosphate |
| CAS | 2392-39-4 |
| Analyte or Component Names | Dexamethasone disodium phosphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 F O8 P . 2 Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dexamethasone Sodium Phosphate, Ak-Dex, Baldex, Colvasone, Corson, Corson (pharmaceutical), Dalalone, Decadron phosphate, Decdan, Dexabene, Dexacort, Dexadreson, Dexagel, Dexagro, Dexamethasone 21-(disodium phosphate), Dexamethasone 21-phosphate disodium salt, Dexamethasone disodium phosphate, Dexamethasone phosphate disodium salt, Dexamethasone phosphate sodium salt, Dexamethasone sodium phosphate, Dexamethazone sodium phosphate, Dezone, Disodium dexamethasone 21-monophosphate, Disodium dexamethasone 21-phosphate, Disodium dexamethasone phosphate, Maxidex Ointment, Megacort, Onadron, Orgadrone, Sodium dexamethasone phosphate, Soldesam, Solu-Deca, Solu-Decadron, Spersadex, Turbinaire, (11beta,16alpha)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione sodium salt (1:2), (11beta,16alpha)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione disodium salt, 9-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt, 9-Fluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione 21-phosphate disodium salt |
| InChI Formula | InChI=1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15+,16+,17+,19+,20+,21+,22+;;/m1../s1 |
| Solution Type | Neat |
| SMILES | [Na+].[Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)([O-])[O-] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction