missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone phosphate disodium, MedChemExpress

Product Code. 16472286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472286 500 mg 500mg
16458816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472286 Supplier Medchem Express Supplier No. HYB1829A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dexamethasone phosphate disodium is a glucocorticoid receptor agonist.

Dexamethasone phosphate disodium is a glucocorticoid receptor agonist.

Specifications

Chemical Name or Material Dexamethasone phosphate disodium
CAS 2392-39-4
Color White
Purity Grade Notes Research
Molecular Formula C22H28FNa2O8P
Quantity 500 mg
Synonym Dexamethasone 21-phosphate disodium salt
Solubility Information H2O : ≥ 100 mg/mL (193.65 mM) ∣DMSO : 1 mg/mL (1.94 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2(F)[C@@H](O)C[C@@]4(C)[C@@]3([H])C[C@@H](C)[C@]4(O)C(COP(O[Na])(O[Na])=O)=O)=C1
Molecular Weight (g/mol) 516.4
Formula Weight 516.4
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.