missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone 9,11-epoxide, MedChemExpress

Product Code. 16498948
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498948 10 mg 10mg
16489056 100 mg 100mg
16464817 50 mg 50mg
16441506 200 mg 200mg
16422376 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498948 Supplier Medchem Express Supplier No. HYN0348
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.

Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.

Specifications

Chemical Name or Material Dexamethasone 9,11-epoxide
CAS 24916-90-3
Color White
Purity Grade Notes Research
Molecular Formula C22H28O5
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (268.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@]3(O5)[C@]5([H])C2)=O
Molecular Weight (g/mol) 372.45
Formula Weight 372.45
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.