missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Dexamethasone

Product Code. 17150022
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17150022 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17150022 Supplier STEMCELL Technologies Supplier No. 72092
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glucocorticoid pathway activator; Activates glucocorticoid receptor

  • Dexamethasone is a synthetic glucocorticoid, similar to the natural glucocorticoid hydrocortisone
  • Dexamethasone has an increased affinity for glucocorticoid receptors when compared to the natural hydrocortisone ligand (Kd = 5 nM vs 17 nM)
  • Promotes transdifferentiation of hepatocytes from mouse pancreatic cells (Shen et al.)
  • Promotes osteogenic, adipogenic, and chondrogenic differentiation of human mesenchymal cells (Jaiswal et al.), Mackay et al.), Pittenger et al.)
  • Promotes osteogenic, adipogenic, and chondrogenic differentiation of mouse mesenchymal cells (Tropel et al.)
  • Promotes differentiation of mature hepatocytes from mouse and human embryonic stem (ES) cells (Cai et al.), Kubo et al.)
  • Promotes maturation of fetal mouse hepatocytes (Kamiya et al.)

Specifications

CAS 50-02-2
Synonym MK 125; NSC 34521
Quantity 500 mg
Molecular Formula C22H29FO5
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.