missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone 11,21-diacetate, TRC

Product Code. 30008324
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008324 500 mg 500mg
30007676 50 mg 50mg
17901335 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008324 Supplier Toronto Research Chemicals Supplier No. TRCD298925500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3949-26-6
Molecular Formula C26 H33 F O7
Molecular Weight (g/mol) 476.53
Synonym Pregna-1,4-diene-3,20-dione, 11,21-bis(acetyloxy)-9-fluoro-17-hydroxy-16-methyl-, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 11,21-diacetate (7CI,8CI), 9-Fluoro-17-hydroxy-16α-methyl-3,20-dioxopregna-1,4-diene-11β,21-diyl Diacetate, Dexamethasone 11,21-Diacetate
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-11-acetyloxy-9-fluoro-17-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@H](C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C)OC(=O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C26H33FO7/c1-14-10-20-19-7-6-17-11-18(30)8-9-23(17,4)25(19,27)22(34-16(3)29)12-24(20,5)26(14,32)21(31)13-33-15(2)28/h8-9,11,14,19-20,22,32H,6-7,10,12-13H2,1-5H3/t14-,19+,20+,22+,23+,24+,25+,26+/m1/s1
Formula Weight 476.221
Chemical Name or Material Dexamethasone 11,21-diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.