missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexamethasone 11,21-Diacetate, Mikromol™
Description
Dexamethasone 11,21-Diacetate
Specifications
Specifications
| Chemical Name or Material | Dexamethasone 11,21-diacetate |
| CAS | 3949-26-6 |
| Analyte or Component Names | Dexamethasone 11,21-diacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H33 F O7 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pregna-1,4-diene-3,20-dione, 11,21-bis(acetyloxy)-9-fluoro-17-hydroxy-16-methyl-, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 11,21-diacetate (7CI,8CI), 9-Fluoro-17-hydroxy-16α-methyl-3,20-dioxopregna-1,4-diene-11β,21-diyl Diacetate, Dexamethasone 11,21-Diacetate, Dexamethasone Acetate Impurity I |
| InChI Formula | InChI=1S/C26H33FO7/c1-14-10-20-19-7-6-17-11-18(30)8-9-23(17,4)25(19,27)22(34-16(3)29)12-24(20,5)26(14,32)21(31)13-33-15(2)28/h8-9,11,14,19-20,22,32H,6-7,10,12-13H2,1-5H3/t14-,19+,20+,22+,23+,24+,25+,26+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@H](C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C)OC(=O)C |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-11-acetyloxy-9-fluoro-17-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?