missing translation for 'onlineSavingsMsg'
Learn More

Devarda'S Alloy Silylation Reagent, Pure, Fisher Chemical

Product Code. 12337143 Shop All Fisher Chemical Products
Change view
Click to view available options
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
12337143 Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12337143 Supplier Fisher Chemical Supplier No. D/0100/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

250g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
CAS 8049-11-4
Melting Point 550°C
Boiling Point 1700°C
Packaging Plastic powder jar
Form Solid
Molecular Formula Al9Cu10Zn
MDL Number 198176
InChI Key MPKKOQSNGWAMJD-UHFFFAOYSA-N
Molecular Weight (g/mol) 943.67
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Devarda's alloy
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.