missing translation for 'onlineSavingsMsg'
Learn More

Deuterium oxide, for NMR, 99.8 atom % D

Product Code. 16130891 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
25 mL
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
10mL
25mL
500mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 16130891

Brand: Thermo Scientific Acros 166305000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7789-20-0 | H2O | 20.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-20-0
Molecular Formula H2O
Molecular Weight (g/mol) 20.03
MDL Number MFCD00044636
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
Synonym heavy water d2o, deuterated water, heavy water-d2, deuterium oxide usan, water sup 2-h2, deuterium oxide, dideuterium oxide, water, heavy d2-o, heavy water, water-d2
PubChem CID 24602
ChEBI CHEBI:41981
IUPAC Name [(²H)oxy](²H)
SMILES [2H]O[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 1.1050 g/mL
Deuterium (D) 99.75% min.
Packaging Glass Bottle
Refractive Index 1.328
Specific Gravity 1.105
Viscosity 1.25 mPa.s (20°C)
Quantity 500 mL
Melting Point 3.8°C
Boiling Point 101.4°C
Linear Formula D2O
Merck Index 15, 2942
Solubility Information Solubility aq. soln.: miscible, exothermic reaction.
Formula Weight 20.02
Grade NMR
Chemical Name or Material Deuterium oxide, 99.8 atom % D
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.