missing translation for 'onlineSavingsMsg'
Learn More

Deuterium chloride, for NMR, 20 wt. % solution in D2O, 99+ atom % D

Product Code. 10605242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass Bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10605242 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10605242 Supplier Thermo Scientific Acros Supplier No. 175420500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7698-05-7 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 1.2000g/mL
Deuterium (D) 99.0% min.
Packaging Glass Bottle
Refractive Index 1.3726
Quantity 50 g
CAS 7698-05-7
Boiling Point 108.0°C
Molecular Formula ClH
Linear Formula DCl
MDL Number MFCD00044642
Synonym deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES Cl
Molecular Weight (g/mol) 36.46
PubChem CID 522634
Formula Weight 37.47
Grade NMR
Chemical Name or Material Deuterium chloride, 99+ atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Deuterium chloride, 20 wt. % solution in D2O
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Deuterium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.