missing translation for 'onlineSavingsMsg'
Learn More

Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic)

Product Code. 11354558
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354558 50 g 50g
11344558 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11354558 Supplier Thermo Scientific Alfa Aesar Supplier No. 042408.18
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

NMR Solvent | CAS: 13536-59-9 | BrH | 81.918 g/mol

NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-59-9
Molecular Formula BrH
Molecular Weight (g/mol) 81.918
MDL Number MFCD00044232
InChI Key CPELXLSAUQHCOX-DYCDLGHISA-N
Synonym deuterium bromide, hydrobromic acid-d, deutero-bromide, dbr solution, deuterium bromide solution, bromo 2 h, deuterium bromide, 99 atom % d, deuterium bromide w/w in d2o, deuterium bromide 98.0atom%d in d2o, deuterium bromide solution, 47 wt. % in d2o, 99 atom % d
PubChem CID 518779
SMILES Br

Specifications

Physical Form Liquid
Color Colorless
Packaging Clear Glass Ampule
Sensitivity Light and Moisture
Quantity 50 g
Boiling Point 126°C
UN Number UN1788
Formula Weight 81.93
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) 48% w/w in D2O
Chemical Name or Material Deuterium bromide, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.