missing translation for 'onlineSavingsMsg'
Learn More

Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic)

Product Code. 11344558
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344558 10 g 10g
11354558 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11344558 Supplier Thermo Scientific Alfa Aesar Supplier No. 042408.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

NMR Solvent | CAS: 13536-59-9 | BrH | 81.918 g/mol

NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13536-59-9
Molecular Formula BrH
Molecular Weight (g/mol) 81.918
MDL Number MFCD00044232
InChI Key CPELXLSAUQHCOX-DYCDLGHISA-N
Synonym deuterium bromide, hydrobromic acid-d, deutero-bromide, dbr solution, deuterium bromide solution, bromo 2 h, deuterium bromide, 99 atom % d, deuterium bromide w/w in d2o, deuterium bromide 98.0atom%d in d2o, deuterium bromide solution, 47 wt. % in d2o, 99 atom % d
PubChem CID 518779
SMILES Br

Specifications

Physical Form Liquid
Color Colorless
Packaging Clear Glass Ampule
Sensitivity Light and Moisture
Quantity 10 g
Boiling Point 126°C
UN Number UN1788
Formula Weight 81.93
Percent Purity ≥99%
Concentration or Composition (by Analyte or Components) 48% w/w in D2O
Chemical Name or Material Deuterium bromide, Isotopic
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Deuterium bromide, 48% w/w in deuterium oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Deuterium bromide

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.