missing translation for 'onlineSavingsMsg'
Learn More

Deuterated Phthalates Mixture 634 1000 μg/mL in Ethyl acetate, Dr. Ehrenstorfer

Product Code. 30318043
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318043 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318043 Supplier Dr. Ehrenstorfer Supplier No. DREA50000634EA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-2-ethylhexyl ester D4∣Phthalic acid, bis-butyl ester D4∣Phthalic acid, bis-ethyl ester D4
CAS 93951-87-2, 93952-11-5, 93952-12-6
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Phthalic acid, bis-2-ethylhexyl ester D4 1000 μg/mL∣Phthalic acid, bis-butyl ester D4 1000 μg/mL∣Phthalic acid, bis-ethyl ester D4 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.