missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Detoxification Buffer ERB

Product Code. 15867606
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867606 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15867606 Supplier Macherey-Nagel™ Supplier No. 740495.1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detoxification Buffer ERB (1000 mL) for transfection-grade Plasmid DNA Isolation

Endotoxins are co-purified during typical plasmid preparations from bacterial lysates. Since they interfere with eukaryotic cell survival, endotoxin reduction is essential prior to cell transfection. NucleoSpin™ Plasmid Transfection-grade kits are designed to reduce endotoxins from bacterial lysates.

  • Purifies high-copy plasmids for transfection
  • Novel technology to diminish endotoxin content (< 50 EU/μg DNA) for successful transfection
  • Rapid purification from E. coli via automatic, manual, vacuum or centrifugation processing
  • Ultra-fast procedure simplifies the work-flow
  • High-throughput — 96-well plates allow for easy automation and supports plasmid screening
  • Downstream applications: transfection of common cell lines (lipofection, electroporation)
  • Also suitable for standard molecular biology application, such as cloning and sequencing

Specifications

Chemical Name or Material Detoxification buffer ERB
CAS 102-71-6
Color Colorless
For Use With (Equipment) NucleoSpin Plasmid/96 Plasmid Transfection-grade full and core kit
pH 5.5 to 6.5
Physical Form Liquid
Quantity 1000 mL
Formulation Triethanolamine; 2-Propanol; Water
Odor Alcoholic
Solubility Information Soluble in Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.