missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Detomidine Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Detomidine hydrochloride |
| CAS | 90038-01-0 |
| Analyte or Component Names | Detomidine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H14 N2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Detomidine hydrochloride, 4-(2,3-Dimethylbenzyl)-1H-imidazole hydrochloride, 1H-Imidazole, 4-[(2,3-dimethylphenyl)methyl]-, monohydrochloride, Detomidine hydrochloride, Domosedan, Dormosedan, MPV 253AII |
| InChI Formula | InChI=1S/C12H14N2.ClH/c1-9-4-3-5-11(10(9)2)6-12-7-13-8-14-12;/h3-5,7-8H,6H2,1-2H3,(H,13,14);1H |
| Solution Type | Neat |
| SMILES | Cl.Cc1cccc(Cc2c[nH]cn2)c1C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction