Learn More
Cole-Parmer™ Botulinum Toxin Detection Kit
Product Code. 13234493 Shop All Cole-Parmer Products
Change view
Click to view available options
For use with: Toxin Detection.
Product Code.
13234493 For use with: Toxin Detection.
1 options
Product Code. 13234493 Supplier Cole-Parmer™ Supplier No. 3610814
Quantity Price
1 £564.00 / Pack of 1
5 £515.78 / Pack of 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Accurately detect small, yet high threat amounts of biowarfare agents-without extensive sample preparation or additional expensive equipment.
  • Results in 3- 30 minutes
  • Easy to read – two lines for positive; one line for negative
  • High Sensitivity and specificity
  • Biohazard disposal bags and chain of custody labels are included for proper control and disposal or transport of the sample and test
Specifications
Description Botulinum Toxin Detection Kit, handheld assay, 10/bx
Test Type Botulinum Toxin
Quantity 10/Box