missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Detection Enhancer, for real-time PCR

Product Code. 30057791 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
100 x 25 μL Reactions
500 x 25 μL Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
30057791 500 x 25 μL Reactions 500 reactions
30054472 100 x 25 μL Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30057791 Supplier Applied Biosystems™ Supplier No. A44941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formulated to improve template amplification in real-time PCR (qPCR) workflows

  • Detection Enhancer is uniquely formulated to improve template amplification in real-time PCR (qPCR) workflows. This reagent is particularly beneficial with DNA samples containing GC-rich sequences and/or persistent secondary structure. Detection Enhancer is a component of the AgPath-ID One-Step RT-qPCR Reagents kit and is offered here as a stand-alone product that can be used with any Applied Biosystem real-time PCR master mix.
  • Improve qPCR amplification of templates containing high GC content and persistent secondary structure
  • Compatible with any Applied Biosystem real-time PCR master mix
  • Flexible kit sizes for a range of laboratory throughputs
  • GC-rich DNA sequences are usually defined as those that contain >60% cytosine (C) or guanine (G). GC-rich samples can be particularly challenging to amplify because the sequences are more stable than those with lower GC content. In qPCR experiments, this results in a higher DNA melting point. In addition, GC-rich sequences can form secondary structure, which can add further amplification challenges. Adding a small volume of Detection Enhancer to your reaction mix can dramatically improve qPCR amplification in GC-rich samples.
  • Detection Enhancer is specifically formulated to address qPCR amplification in particularly stable DNA sequences. It is only for use in real-time PCR workflows where amplification is affected by templates that are GC rich and/or contain persistent secondary structure. The reagents may affect sensitivity for other target types, so it is recommended to first assemble the reaction without Detection Enhancer. If no signal is observed from samples that are expected to be positive, then add Detection Enhancer to the reaction.
  • The recommended volume of Detection Enhancer per 25 μL reaction is 1.67 μL. For more information on applications of Detection Enhancer, please see the AgPath-ID One-Step RT-qPCR Reagents kit user guide
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 1.2 mL Detection Enhancer, store at -5 to -30°C
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 System
Product Line Applied Biosystems
Product Type Detection Enhancer
Quantity 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 500 x 25 μL Reactions
Sample Type Viral Samples
Volume 1.2 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.