missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Detection Enhancer, for real-time PCR
Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
100 x 25 μL Reactions
500 x 25 μL Reactions
Unit Size:
100 reactions
500 reactions
Description
- Detection Enhancer is uniquely formulated to improve template amplification in real-time PCR (qPCR) workflows. This reagent is particularly beneficial with DNA samples containing GC-rich sequences and/or persistent secondary structure. Detection Enhancer is a component of the AgPath-ID One-Step RT-qPCR Reagents kit and is offered here as a stand-alone product that can be used with any Applied Biosystem real-time PCR master mix.
- Improve qPCR amplification of templates containing high GC content and persistent secondary structure
- Compatible with any Applied Biosystem real-time PCR master mix
- Flexible kit sizes for a range of laboratory throughputs
- GC-rich DNA sequences are usually defined as those that contain >60% cytosine (C) or guanine (G). GC-rich samples can be particularly challenging to amplify because the sequences are more stable than those with lower GC content. In qPCR experiments, this results in a higher DNA melting point. In addition, GC-rich sequences can form secondary structure, which can add further amplification challenges. Adding a small volume of Detection Enhancer to your reaction mix can dramatically improve qPCR amplification in GC-rich samples.
- Detection Enhancer is specifically formulated to address qPCR amplification in particularly stable DNA sequences. It is only for use in real-time PCR workflows where amplification is affected by templates that are GC rich and/or contain persistent secondary structure. The reagents may affect sensitivity for other target types, so it is recommended to first assemble the reaction without Detection Enhancer. If no signal is observed from samples that are expected to be positive, then add Detection Enhancer to the reaction.
- The recommended volume of Detection Enhancer per 25 μL reaction is 1.67 μL. For more information on applications of Detection Enhancer, please see the AgPath-ID One-Step RT-qPCR Reagents kit user guide
Specifications
Specifications
| Content And Storage | 1 x 190 μL Detection Enhancer, store at -5 to -30°C |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | High |
| PCR Method | 1-step RT-qPCR |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7500 System |
| Product Line | Applied Biosystems |
| Product Type | Detection Enhancer |
| Quantity | 100 reactions |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction