missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DET1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546710
This item is not returnable.
View return policy
Description
Peptide sequence: DCRCVIVGSA AYLPDEPHPP FFEVYRNSES VTPNPRSPLE DYSLHIIDLH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
DET1 is a component of the E3 ubiquitin ligase DCX DET1-COP1 complex, which is required for ubiquitination and subsequent degradation of target proteins. The complex is involved in JUN ubiquitination and degradation.
Specifications
| DET1 | |
| Polyclonal | |
| Unconjugated | |
| DET1 | |
| 2610034H20Rik; de-etiolated 1; de-etiolated homolog 1; de-etiolated homolog 1 (Arabidopsis); de-etiolated-1 homolog; DET1; DET1 homolog; DET1 partner of COP1 E3 ubiquitin ligase; DET1, COP1 ubiquitin ligase partner; RGD1309793; wu:fc18d06; zgc:66486 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55070 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7L5Y6 | |
| DET1 | |
| Synthetic peptide directed towards the N-terminal of human DET1 (aa 150-199). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction