missing translation for 'onlineSavingsMsg'
Learn More

Desvenlafaxine, 97%

Product Code. 15678745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15678745 5 g 5g
15668745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15678745 Supplier Thermo Scientific Acros Supplier No. 461570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93413-62-8 | C16H25NO2 | 263.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93413-62-8
Molecular Formula C16H25NO2
Molecular Weight (g/mol) 263.38
InChI Key KYYIDSXMWOZKMP-UHFFFAOYSA-N
Synonym desvenlafaxine, o-desmethylvenlafaxine, 4-2-dimethylamino-1-1-hydroxycyclohexyl ethyl phenol, pristiq, o-desmethyl venlafaxine, desvenlafaxine inn:ban, desvenlafaxine inn, o-desmethylvenlafaxine odv, 1-2-dimethylamino-1-4-hydroxyphenyl ethyl cyclohexanol
PubChem CID 125017
ChEBI CHEBI:83527
IUPAC Name 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
SMILES CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O

Specifications

Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 263.38
Percent Purity 97%
Chemical Name or Material Desvenlafaxine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.