missing translation for 'onlineSavingsMsg'
Learn More

Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one), LoGiCal

Product Code. 30460883
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460883 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460883 Supplier LoGiCal Supplier No. LGCFOR1569.01
 Restricted Item
Estimated Shipment: 30-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one)
CAS 224789-21-3
Analyte or Component Names Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one)
Shipping Condition Room Temperature
Molecular Formula C17 H20 N4 O2
Certifications/Compliance ISO 9001
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-(2-ethoxyphenyl)-5-methyl-7-propyl-, 2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
InChI Formula InChI=1S/C17H20N4O2/c1-4-8-14-18-11(3)15-17(22)19-16(20-21(14)15)12-9-6-7-10-13(12)23-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H,19,20,22)
Solution Type Neat
SMILES CCCc1nc(C)c2C(=O)NC(=Nn12)c3ccccc3OCC
IUPAC Name 2-(2-ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 312.37
Quantity 10 mg
Formula Weight 312.1586 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.