missing translation for 'onlineSavingsMsg'
Learn More

Desthiazolylmethyl Ritonavir, TRC

Product Code. 30135713
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135713 10 mg 10mg
30004013 50 mg 50mg
30003717 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135713 Supplier Toronto Research Chemicals Supplier No. TRCD29745010MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256328-82-2
Molecular Formula C33 H43 N5 O4 S
Molecular Weight (g/mol) 605.79
Synonym Butanamide, 3-methyl-2-[[[methyl[[2-(1-methylethyl)-4-thiazolyl]methyl]amino]carbonyl]amino]-N-[(1S)-1-[[(4S,5S)-2-oxo-4-(phenylmethyl)-5-oxazolidinyl]methyl]-2-phenylethyl]-, (2S)-, Ritonavir Imp. L (EP), (4S,5S)-4-Benzyl-5-[(2S)-2-[[(2S)-3-methyl-2-[[methyl[[2-(1-methylethyl)thiazol-4-yl]methyl]carbamoyl]-amino]butanoyl]amino]-3-phenylpropyl]oxazolidin-2-one, Desthiazolylmethyl Ritonavir
IUPAC Name (2S)-N-[(2S)-1-[(4S,5S)-4-benzyl-2-oxo-1,3-oxazolidin-5-yl]-3-phenylpropan-2-yl]-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanamide
SMILES CC(C)[C@H](NC(=O)N(C)Cc1csc(n1)C(C)C)C(=O)N[C@H](C[C@@H]2OC(=O)N[C@H]2Cc3ccccc3)Cc4ccccc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C33H43N5O4S/c1-21(2)29(37-32(40)38(5)19-26-20-43-31(35-26)22(3)4)30(39)34-25(16-23-12-8-6-9-13-23)18-28-27(36-33(41)42-28)17-24-14-10-7-11-15-24/h6-15,20-22,25,27-29H,16-19H2,1-5H3,(H,34,39)(H,36,41)(H,37,40)/t25-,27-,28-,29-/m0/s1
Formula Weight 605.3036
Chemical Name or Material Desthiazolylmethyl ritonavir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.