missing translation for 'onlineSavingsMsg'
Learn More

Desoxycarbadox, TRC

Product Code. 30132531
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132531 25 mg 25mg
30003245 100 mg 100mg
30010474 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132531 Supplier Toronto Research Chemicals Supplier No. TRCD29699525MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55456-55-8
Molecular Formula C11 H10 N4 O2
Molecular Weight (g/mol) 230.2227
Synonym Hydrazinecarboxylic acid, (2-quinoxalinylmethylene)-, methyl ester (9CI), Desoxycarbadox, Methyl 3-(2-quinoxalinylmethylene)carbazate
IUPAC Name methyl N-[(E)-quinoxalin-2-ylmethylideneamino]carbamate
SMILES COC(=O)N\N=C\c1cnc2ccccc2n1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H10N4O2/c1-17-11(16)15-13-7-8-6-12-9-4-2-3-5-10(9)14-8/h2-7H,1H3,(H,15,16)/b13-7+
Formula Weight 230.0804
Percent Purity >95%
Chemical Name or Material Desoxycarbadox
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.