missing translation for 'onlineSavingsMsg'
Learn More

Desmoglein-4 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30736056 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736056 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736056 Supplier Novus Biologicals Supplier No. AF6589JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Desmoglein-4 Polyclonal antibody specifically detects Desmoglein-4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein-4
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias Cadherin family member 13, CDGF13, CDHF13CDH family member 13, desmoglein 4, LAHdesmoglein-4
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Desmoglein-4, Glu24-Ala631, Accession # Q86SJ6
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 147409
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.