Learn More
Invitrogen™ Desmoglein 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121194
Description
Positive test controls include: HeLa, Mouse liver. The target is usually found in the following locations: Cell junction, Cell membrane, Single-pass type I membrane protein, desmosome. Immunogen sequence: FGAATGVGIC SSGQSGTMRT RHSTGGTNKD YADGAISMNF LDSYFSQKAF ACAEEDDGQE ANDCLLIYDN EGADATGSPV GSVGCCSFIA DDLDDSFLDS LGPKFKKLAE ISLGVDGEGK EVQPPSKDSG YGIESCGHPI EVQQTGFVKC QTLSGSQGAS ALSTSGSVQP AVSIPDPLQH GNYLVTETYS ASGSLVQPST AGFDPLLTQN VIVTERVICP ISSVPGNLAG PTQLRGSHTM LCTEDPCSRL I.
Desmoglein 3 (Dsg3), also known as Cadherin family member 6 (CDHF6), is a member of the desmosomal cadherin family and plays a critical role in cell-cell adhesion. It is predominately expressed in stratified squamous epithelium, including the skin, oral mucosa, and esophagus. Desmoglein 3 has been characterized as the auto-antigen of pemphigus vulgaris (PV), a life-threatening autoimmune disease. Binding of anti-Desmoglein 3 autoantibodies causes the loss of cell-cell adhesion, which clinically manifests as blisters on the skin and oral mucosa, and histologically as suprabasilar acantholysis.
Specifications
| Desmoglein 3 | |
| Polyclonal | |
| Unconjugated | |
| Dsg3 | |
| 130 kDa pemphigus vulgaris antigen; 130 kDa pemphigus vulgaris antigen homolog; bal; balding; Cadherin family member 6; CDHF6; desmoglein 3; desmoglein-3; DKFZp686P23184; Dsg3; PVA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1830 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32926 | |
| Dsg3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 749-999 of human Desmoglein 3 (NP_001935.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.