missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmoglein 3 Monoclonal Antibody (5G11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516025
This item is not returnable.
View return policy
Description
Desmoglein 3 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow, IP
Desmoglein 3 (Dsg3), also known as Cadherin family member 6 (CDHF6), is a member of the desmosomal cadherin family and plays a critical role in cell-cell adhesion. It is predominately expressed in stratified squamous epithelium, including the skin, oral mucosa, and esophagus. Desmoglein 3 has been characterized as the auto-antigen of pemphigus vulgaris (PV), a life-threatening autoimmune disease. Binding of anti-Desmoglein 3 autoantibodies causes the loss of cell-cell adhesion, which clinically manifests as blisters on the skin and oral mucosa, and histologically as suprabasilar acantholysis.
Specifications
| Desmoglein 3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P32926 | |
| Dsg3 | |
| Human Desmoglein 3 recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 5G11 | |
| Unconjugated | |
| Dsg3 | |
| 130 kDa pemphigus vulgaris antigen; 130 kDa pemphigus vulgaris antigen homolog; bal; balding; Cadherin family member 6; CDHF6; desmoglein 3; desmoglein-3; DKFZp686P23184; Dsg3; PVA | |
| Mouse | |
| Protein G | |
| RUO | |
| 1830 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction