missing translation for 'onlineSavingsMsg'
Learn More

Desmoglein-3 Antibody (DSG3/8613R), Novus Biologicals™

Rabbit Monoclonal Antibody

£205.00 - £377.00

Specifications

Antigen Desmoglein-3
Clone DSG3/8613R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18630115
View Documents
Novus Biologicals
NBP3-23751-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18687944
View Documents
Novus Biologicals
NBP3-23751-20ug
20 μg
£205.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Desmoglein-3 Monoclonal antibody specifically detects Desmoglein-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Desmoglein-3
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction
10 mM PBS with 0.05% BSA
1830
IgG κ
Protein A or G purified
DSG3/8613R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA
Recombinant fragment (around aa300-500) of human Desmoglein-3 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.