missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Antibody (DSG3/8251R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23742-100ug
This item is not returnable.
View return policy
Description
Desmoglein-3 Monoclonal antibody specifically detects Desmoglein-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Desmoglein-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
| Recombinant fragment (around aa300-500) of human Desmoglein-3 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
| 1830 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| DSG3/8251R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction