missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Antibody (5H10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP1-78984
This item is not returnable.
View return policy
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot.
Specifications
| Desmoglein-3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 15588331) | |
| Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
| Mouse | |
| 130 kDa | |
| 0.1 mL | |
| Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
| 1830 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 5H10 | |
| Unconjugated | |
| P32926 | |
| DSG3 | |
| The extracellular domain of human Desmoglein 3 [Swiss-Prot# P32926] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction