missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Desmoglein 2 Recombinant Human Monoclonal Antibody (22H6)
GREENER_CHOICE

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA550212

Product Code. 17928862

  • £367.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Desmoglein 2 Recombinant Monoclonal Antibody for ELISA

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Desmoglein 2
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Q14126
DSG2
Recombinant Human DSG2 protein.
100 μL
Primary
Human
Antibody
IgG1
ELISA, Neutralization
22H6
Unconjugated
DSG2
AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037
Human
Affinity chromatography
RUO
1829
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.