missing translation for 'onlineSavingsMsg'
Learn More

Desmoglein-2 Rabbit anti-Human, Clone: 8O0R6, Novus Biologicals™

Product Code. 18394602 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394602 100 μg 100µL
18344352 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18394602 Supplier Novus Biologicals Supplier No. NBP316421100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Desmoglein-2 Monoclonal antibody specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein-2
Applications Western Blot
Classification Monoclonal
Clone 8O0R6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ARVC10, ARVD10, Cadherin family member 5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, HDGC, MGC117036, MGC117037
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1001-1118 of human Desmoglein-2 (Q14126).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1829
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.