missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmoglein 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569369
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 93%; Rabbit: 85%; Rat: 100%. Synthetic peptide located within the following region: TDADEVGSDN WLANFTFASG NEGGYFHIET DTQTNEGIVT LVKEVDYEEM.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
Specifications
| Desmoglein 2 | |
| Polyclonal | |
| Unconjugated | |
| DSG2 | |
| AA408168; ARVC10; ARVD10; Cadherin family member 5; CDHF5; CMD1BB; D18Ertd293e; desmoglein 2; desmoglein-2; DSG2; DSG-2; HDGC; MGC117034; MGC117036; MGC117037 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13511 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O55111 | |
| DSG2 | |
| synthetic peptide directed towards the middle region of mouse Desmoglein 2. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction