missing translation for 'onlineSavingsMsg'
Learn More

Desmoglein-2 Antibody (6D8), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18423793 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423793 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423793 Supplier Novus Biologicals Supplier No. NBP254530AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Desmoglein-2 Monoclonal antibody specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoglein-2
Applications Western Blot, Flow Cytometry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 6D8
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias ARVC10, ARVD10, Cadherin family member 5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, HDGC, MGC117036, MGC117037
Host Species Mouse
Immunogen Recombinant human Desmoglein-2 protein (Uniprot: Q14126)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1829
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.