missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-1 Antibody (DSG1/1733), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54377AF405
This item is not returnable.
View return policy
Description
Desmoglein-1 Monoclonal antibody specifically detects Desmoglein-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Desmoglein-1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1 | |
| Recombinant human Desmoglein-1 protein fragment corresponding to intracellular domain (Uniprot: Q02413) | |
| 100 μg | |
| Cancer, Cellular Signaling | |
| 1828 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DSG1/1733 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction