missing translation for 'onlineSavingsMsg'
Learn More

Desmocollin-2 Antibody (004), DyLight 350, Novus Biologicals™

Product Code. 18418487 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418487 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418487 Supplier Novus Biologicals Supplier No. NBP289651UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Desmocollin-2 Monoclonal antibody specifically detects Desmocollin-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmocollin-2
Applications ELISA
Classification Monoclonal
Clone 004
Conjugate DyLight 350
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ARVD11, Cadherin family member 2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC3DKFZp686I11137
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Desmocollin-2 (Q02487-1; Met1-Arg684).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1824
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.