missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmocollin-1 Antibody (JE66-57), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32264
This item is not returnable.
View return policy
Description
Desmocollin-1 Monoclonal antibody specifically detects Desmocollin-1 in Mouse,Rat samples. It is validated for Western Blot
Specifications
| Desmocollin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, desmosomal glycoprotein 2/3, DG2/DG3 | |
| Synthetic peptide within Human Desmocollin-1 aa 795-894/ 894. (Uniprot: Q08554) | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| JE66-57 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1823 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction