missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Rabbit anti-Human, Rat, Chicken, Hamster, Clone: DES/2960R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07640-100ug
This item is not returnable.
View return policy
Description
Desmin Monoclonal specifically detects Desmin in Human, Rat, Chicken, Hamster samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Desmin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Recombinant human full-length Desmin protein | |
| 100 μg | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 1674 | |
| Store at 4°C. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DES/2960R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Chicken, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction