Learn More
Epredia™ Lab Vision™ Desmin (Muscle Cell Marker) Ab-1, Mouse Monoclonal Antibody
Specifically detect desmin in human, baboon, monkey, cow, cat, dog, hamster, rat and chicken samples.
Brand: Epredia™ MS-376-R7
Description
Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin. It reacts with leiomyosarcomas (smooth muscle) as well as rhabdomyosarcomas (striated muscle).Host Species: Mouse
Clone: D33
Isotype: IgG1/κ
Species Reactivity: Human, Baboon, Monkey, Cow, Cat, Dog, Hamster, Rat and Chicken. Others not known.
Epitope: Not determined
Immunogen: Desmin from human muscle
Molecular Weight: 53 kDa
Positive Control: Muscle or Sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
| Desmin (Muscle Cell Marker) Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Desmin from human muscle | |
| IVD | |
| Primary | |
| Prediluted |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| D33 | |
| Mouse | |
| 7 mL | |
| Muscle Biology | |
| Bovine, Canine, Chicken, Feline, Hamster, Human, Non-human Primate, Rat | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.