Learn More
Desmin Monoclonal Antibody (DE-U-10), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9747-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Desmin (∽53 kDa) exhibits a high degree of tissue specificity, its expression being predominantly confined to all types of muscle cells (cardiac, skeletal and smooth muscle). Regulation of desmin expression is stage and tissue-specific, since it is induced during terminal development of, for example, skeletal muscle cell differentiation. In skeletal en cardiac muscle cells desmin is localized in the Z-disk region and at the intercalated disk. The expression pattern of desmin in smooth muscle is much more heterogenous. Coexpression of vimentin and desmin has been observed in tumors derived from muscle tissue, i.e. rhabdomyosarcomas and leiomyosarcomas. Furthermore, during myocard dysfunction dramatic changes in the distribution of desmin have been observed.
Desmin (approximately 53 kDa) exhibits a high degree of tissue specificity, its expression being predominantly confined to all types of muscle cells (cardiac, skeletal and smooth muscle). Regulation of desmin expression is stage and tissue-specific, since it is induced during terminal development of, for example, skeletal muscle cell differentiation. In skeletal en cardiac muscle cells desmin is localized in the Z-disk region and at the intercalated disk. The expression pattern of desmin in smooth muscle is much more heterogenous. Coexpression of vimentin and desmin has been observed in tumors derived from muscle tissue, i.e. rhabdomyosarcomas and leiomyosarcomas. Furthermore, during myocard dysfunction dramatic changes in the distribution of desmin have been observed.Specifications
Desmin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
O62654, P02540, P02542, P17661, P31001, P48675 | |
DES | |
Affinity chromatography | |
RUO | |
100328890, 100764990, 101097010, 101117308, 102180712, 13346, 1674, 280765, 395906, 396725, 64362 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DE-U-10 | |
eFluor 660 | |
DES | |
CMD1I; CSM1; CSM2; DES; Desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79_018932; intermediate filament protein; LGMD2R; MFM1; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E; SCPNK; similar to desmin | |
Mouse | |
25 μg | |
Primary | |
Bovine, Rat, Feline, Feline, Hamster, Hamster, Mouse, Mouse, Ovine, Rabbit, Rabbit | |
Antibody | |
IgG1 κ |
For Research Use Only.