missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Antibody (DES/1711), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54503JF549
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human, Mouse, Rat, Chicken, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Desmin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Recombinant human desmin protein (Uniprot: P17661) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 1674 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DES/1711 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction