missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Antibody (CL4501) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59055AF594
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Desmin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| This antibody was developed using a synthetic peptide derived from P17661, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 1674 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL4501 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction