missing translation for 'onlineSavingsMsg'
Learn More

Desmethyltriflupromazine Hydrochloride (Nortriflupromazine Hydrochloride), Mikromol™

Product Code. 30049523
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049523 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049523 Supplier Mikromol Supplier No. MM1675.070025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Desmethyltriflupromazine Hydrochloride (Nortriflupromazine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethyltriflupromazine Hydrochloride
CAS 10157-71-8
Analyte or Component Names Desmethyltriflupromazine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H17 F3 N2 S . Cl H
Certifications/Compliance ISO 9001
Synonym N-Methyl-2-(trifluoromethyl)-10H-phenothiazine-10-propanamine Hydrochloride (1:1), 10H-Phenothiazine-10-propanamine, N-methyl-2-(trifluoromethyl)-, monohydrochloride (9CI), Phenothiazine, 10-[3-(methylamino)propyl]-2-(trifluoromethyl)-, hydrochloride (7CI,8CI), Desmethyltriflupromazine Hydrochloride
InChI Formula InChI=1S/C17H17F3N2S.ClH/c1-21-9-4-10-22-13-5-2-3-6-15(13)23-16-8-7-12(11-14(16)22)17(18,19)20;/h2-3,5-8,11,21H,4,9-10H2,1H3;1H
SMILES Cl.CNCCCN1c2ccccc2Sc3ccc(cc13)C(F)(F)F
IUPAC Name N-methyl-3-[2-(trifluoromethyl)phenothiazin-10-yl]propan-1-amine;hydrochloride
Molecular Weight (g/mol) 374.85
Quantity 25 mg
Formula Weight 374.0831 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.