missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethyltadalafil (Nortadalafil), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Desmethyltadalafil (Nortadalafil)
Specifications
Specifications
| Chemical Name or Material | N-Desmethyl Tadalafil |
| CAS | 171596-36-4 |
| Analyte or Component Names | N-Desmethyl Tadalafil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H17 N3 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-, (6R,12aR)-, Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-, (6R-trans)-, Nortadalafil |
| InChI Formula | InChI=1S/C21H17N3O4/c25-18-9-22-21(26)15-8-13-12-3-1-2-4-14(12)23-19(13)20(24(15)18)11-5-6-16-17(7-11)28-10-27-16/h1-7,15,20,23H,8-10H2,(H,22,26)/t15-,20-/m1/s1 |
| SMILES | O=C1CNC(=O)[C@H]2Cc3c([nH]c4ccccc34)[C@H](N12)c5ccc6OCOc6c5 |
| IUPAC Name | (2R,8R)-2-(1,3-benzodioxol-5-yl)-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction