missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethylfluvoxamine, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Desmethylfluvoxamine
Specifications
Specifications
| Chemical Name or Material | Desmethyl Fluvoxamine |
| CAS | 192876-02-1 |
| Analyte or Component Names | Desmethyl Fluvoxamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H19 F3 N2 O2 |
| Synonym | (1 E)-5-hydroxy-1-[4-(trifluoromethyl)phenyl]-1-Pentanone O-(2-aminoethyl)oxime, 1-Pentanone, 5-hydroxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (E)-, Fluvoxamino acid, Fluvoxamine Maleate Imp. G (EP) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | NCCO\N=C(/CCCCO)\c1ccc(cc1)C(F)(F)F |
| IUPAC Name | (5 E)-5-(2-aminoethoxyimino)-5-[4-(trifluoromethyl)phenyl]pentan-1-ol |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction