missing translation for 'onlineSavingsMsg'
Learn More

Desmethylclozapine (Norclozapine) 1.0 mg/mL in Methanol, LoGiCal

Product Code. 17990530
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17990530 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17990530 Supplier LoGiCal Supplier No. LGCAMP0480.0301
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1 mL

Desmethylclozapine (Norclozapine) 1.0 mg/mL in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethylclozapine
CAS 6104-71-8
Analyte or Component Names N-Desmethylclozapine
Shipping Condition Room Temperature
Molecular Formula C17 H17 Cl N4
Certifications/Compliance ISO 17034
Synonym 8-Chloro-11-(1-piperazinyl)-5H-dibenzo[B,E][1,4]diazepine, Clozapine Imp. C (EP), 8-Chloro-11-(piperazin-1-yl)-5H-dibenzo[b,e][1,4]diazepine, Desmethylclozapine, Norclozapine, Clozapine Impurity C
InChI Formula InChI=1S/C17H17ClN4/c18-12-5-6-15-16(11-12)21-17(22-9-7-19-8-10-22)13-3-1-2-4-14(13)20-15/h1-6,11,19-20H,7-10H2
SMILES Clc1ccc2Nc3ccccc3C(=Nc2c1)N4CCNCC4
IUPAC Name 3-chloro-6-piperazin-1-yl-11H-benzo[b][1,4]benzodiazepine
Solvent or Matrix Methanol
Molecular Weight (g/mol) 312.8
Quantity 1 mL
Formula Weight 312.1142 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.