missing translation for 'onlineSavingsMsg'
Learn More

Desmethylcitalopram Hydrochloride, LoGiCal

Product Code. 30463894
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463894 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463894 Supplier LoGiCal Supplier No. LGCFOR0242.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethylcitalopram Hydrochloride
CAS 97743-99-2
Analyte or Component Names Desmethylcitalopram Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H19 F N2 O . Cl H
Certifications/Compliance ISO 9001
Synonym Citalopram Hydrobromide Imp. D (EP), Citalopram Imp. D (EP), Citalopram USP Related Compound D, Citalopram USP RC D, (1RS)-1-(4-Fluorophenyl)-1-[3-(methylamino)propyl]-1,3-dihydroisobenzofuran-5-carbonitrile Hydrochloride, Citalopram Hydrobromide Imp. D (EP) as Hydrochloride, Desmethylcitalopram Hydrochloride, 1-(4-Fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-5-isobenzofurancarbonitrile monohydrochloride, Lu 11-109C, Citalopram Related Compound D as Hydrochloride, Citalopram Hydrobromide Impurity D as Hydrochloride, Escitalopram Oxalate Impurity D as Hydrochloride, Citalopram Impurity D as Hydrochloride, Escitalopram Impurity D as Hydrochloride
InChI Formula InChI=1S/C19H19FN2O.ClH/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;/h3-8,11,22H,2,9-10,13H2,1H3;1H
Solution Type Neat
SMILES Cl.CNCCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3
IUPAC Name 1-(4-fluorophenyl)-1-[3-(methylamino)propyl]-3H-2-benzofuran-5-carbonitrile;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 346.83
Quantity 10 mg
Formula Weight 346.1248 g/mol
Show More Show Less
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.