missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethylchlorphenamine Maleate 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Desmethylchlorphenamine Maleate 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Desmethylchlorphenamine Maleate |
| CAS | 22630-25-7 |
| Analyte or Component Names | Desmethylchlorphenamine Maleate |
| Shipping Condition | Ice Pack (-20°C) |
| Molecular Formula | C15 H17 Cl N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | (3 RS)-3-(4-Chloro-phenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine Maleate, Chlorphenamine Maleate Imp. C (EP) as Maleate, Desmethylchlorphenamine Maleate, Chlorphenamine Maleate Imp. C (EP), Chlorphenamine Imp. C (EP), Chlorpheniramine USP Related Compound C, Chlorpheniramine USP RC C, Chlorpheniramine Related Compound C as Maleate, Chlorphenamine Maleate Impurity C as Maleate, Chlorphenamine Impurity C as Maleate |
| InChI Formula | InChI=1 S/C15H17ClN2.C4H4O4/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12;5-3(6)1-2-4(7)8/h2-8,10,14,17 H,9,11H2,1H3;1-2 H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CNCCC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-N-methyl-3-pyridin-2-ylpropan-1-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction