missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Levofloxacin, TRC

Product Code. 30011195
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011195 5 mg 5mg
30010623 10 mg 10mg
30003978 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011195 Supplier Toronto Research Chemicals Supplier No. TRCD2918445MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117707-40-1
Molecular Formula C17 H18 F N3 O4
Molecular Weight (g/mol) 347.34
Synonym 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-, (3S)-, 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-, (S)-, (3S)-9-Fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, DN 5455, Desmethyl Levofloxacin, UING 4-255, (S)-9-Fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, Levofloxacin Rel. Comp. A (USP), N-Desmethyllevofloxacin
IUPAC Name (2S)-7-fluoro-2-methyl-10-oxo-6-piperazin-1-yl-4-oxa-1-azatricyclo[7.3.1.05,13]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
SMILES C[C@H]1COc2c(N3CCNCC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H18FN3O4/c1-9-8-25-16-13-10(15(22)11(17(23)24)7-21(9)13)6-12(18)14(16)20-4-2-19-3-5-20/h6-7,9,19H,2-5,8H2,1H3,(H,23,24)/t9-/m0/s1
Formula Weight 347.1281
Percent Purity >95%
Chemical Name or Material N-Desmethyllevofloxacin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.