missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Atomoxetine Hydrochloride, TRC

Codice prodotto. 30136622
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136622 10 mg 10mg
30133588 1 mg 1mg
18012440 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136622 Supplier Toronto Research Chemicals Supplier No. TRCD29082510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881995-46-6
Molecular Formula C16H20ClNO
Molecular Weight (g/mol) 277.79
Synonym rac-N-Desmethyl Atomoxetine HCl
IUPAC Name 3-phenyl-3-(o-tolyloxy)propan-1-amine hydrochloride
SMILES Cl.Cc1ccccc1O[C@H](CCN)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H19NO.ClH/c1-13-7-5-6-10-15(13)18-16(11-12-17)14-8-3-2-4-9-14;/h2-10,16H,11-12,17H2,1H3;1H
Formula Weight 277.12
Percent Purity >95%
Chemical Name or Material Desmethyl Atomoxetine HCl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.