missing translation for 'onlineSavingsMsg'
Learn More

Deslorelin Acetate, TRC

Product Code. 30138215
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138215 25 mg 25mg
30007467 5 mg 5mg
30006027 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138215 Supplier Toronto Research Chemicals Supplier No. TRCD29029025MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1642579-30-3
Synonym Deslorelin acetate∣1-9-Luteinizing hormone-releasing factor (swine), 6-D-tryptophan-9-(N-ethyl-L-prolinamide)-, acetate (1:?)∣6-D-Tryptophan-9-(N-ethyl-L-prolinamide)-10-deglycinamide-luteinizing hormone-releasing factor (pig) acetate (salt)∣6-D-Trypto

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 1342.5
Product Type Metabolite
Quantity 25 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.