missing translation for 'onlineSavingsMsg'
Learn More

Desloratadine, MedChemExpress

Product Code. 16498356
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498356 100 mg 100mg
16482166 500 mg 500mg
16404397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498356 Supplier Medchem Express Supplier No. HYB0539
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.

Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.

Specifications

Chemical Name or Material Desloratadine
Physical Form Solid
CAS 100643-71-8
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H19ClN2
For Use With (Application) COVID-19-immunoregulation
Synonym Sch34117
Solubility Information DMSO : 25 mg/mL (80.43 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.16 mM; Need ultrasonic)
SMILES ClC1=CC=C2C(CCC3=CC=CN=C3/C2=C4CCNCC/4)=C1
Molecular Weight (g/mol) 310.82
Formula Weight 310.82
Percent Purity 99.98%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.